Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

34053

Supelco

Atrazine-d5

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-4-pentadeuteroethylamino-6-isopropylamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9D5ClN5
CAS Number:
Molecular Weight:
220.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC(C)C)n1

InChI

1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)/i1D3,4D2

InChI key

MXWJVTOOROXGIU-SGEUAGPISA-N

General description

Atrazine-d5 is a deuterated labeled isotope of the triazine pesticide, atrazine.

Application

Atrazine-d5 may be used as an analytical standard for the
determination of atrazine in water and soil samples using isotope dilution gas chromatography coupled with mass spectrometry.
Atrazine-d5 may also be used as an internal standard for the estimation of atrazine in water samples using solid-phase extraction and polar organic chemical integrative samplers (POCIS) followed by liquid chromatography with electrospray ionization-tandem mass spectrometry.(LC-ESI-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Heart

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianglin Peng et al.
Chemosphere, 266, 128933-128933 (2020-11-24)
As a macrolide antibiotic, clarithromycin (CLA) has a high detection rate in surface water and sewage treatment plant effluents worldwide, posing a considerably high ecological risk to aquatic ecosystem. However, algal transcriptome and metabolome in response to CLA remains largely
Jiahua Guo et al.
Ecotoxicology and environmental safety, 201, 110737-110737 (2020-06-09)
Macrolide antibiotics are common contaminants in the aquatic environment. They are toxic to a wide range of primary producers, inhibiting the algal growth and further hindering the delivery of several ecosystem services. Yet the molecular mechanisms of macrolides in algae
Determination of atrazine, lindane, pentachlorophenol, and diazinon in water and soil by isotope dilution gas chromatography/mass spectrometry.
Lopez-Avila V, et al.
Analytical Chemistry, 57(14), 2797-2801 (1985)
Liquid chromatography coupled with tandem mass spectrometry method for thirty-three pesticides in natural water and comparison of performance between classical solid phase extraction and passive sampling approaches
Lissalde S, et al.
Journal of Chromatography A, 1218(11), 1492-1502 (2011)
Sophie Lissalde et al.
The Science of the total environment, 488-489, 188-196 (2014-05-17)
This study focuses on how Polar Organic Chemical Integrative Samplers (POCIS) work in real environmental conditions. A selection of 23 polar pesticides and 8 metabolites were investigated by exposure of triplicates of integrative samplers in two rivers in France for

Articles

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service