Skip to Content
MilliporeSigma
All Photos(1)

Documents

33762

Supelco

Microcystin-LF solution

6-9 μg/mL in methanol, analytical standard

Synonym(s):

Algae bloom toxin, Biotoxin, Algae bloom standard, Cyanobacterial toxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H71N7O12
CAS Number:
Molecular Weight:
986.16
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

6-9 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@@H](Cc1ccccc1)[C@@H](C)\C=C(C)\C=C\[C@@H]2NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@@H](C)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](C)NC(=O)C(=C)N(C)C(=O)CC[C@@H](NC(=O)[C@H]2C)C(O)=O)C(O)=O

InChI

1S/C52H71N7O12/c1-29(2)25-40-50(66)58-44(52(69)70)33(6)46(62)56-41(27-36-17-13-11-14-18-36)49(65)54-38(22-21-30(3)26-31(4)42(71-10)28-37-19-15-12-16-20-37)32(5)45(61)55-39(51(67)68)23-24-43(60)59(9)35(8)48(64)53-34(7)47(63)57-40/h11-22,26,29,31-34,38-42,44H,8,23-25,27-28H2,1-7,9-10H3,(H,53,64)(H,54,65)(H,55,61)(H,56,62)(H,57,63)(H,58,66)(H,67,68)(H,69,70)/b22-21+,30-26+/t31-,32-,33-,34+,38-,39+,40-,41-,42-,44+/m0/s1

InChI key

FEVBMCJUKWWWBT-QVWKUIOOSA-N

General description

Microcystin-LF is a toxic analog of microcystin produced by cyanobacteria in freshwater systems.

Application

Microcystin-LF may be used as an analytical reference standard for the determination of the analyte in raw and treated waters using high-performance liquid chromatography with photo-diode array detection (HPLC-PDA).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Extraction and high-performance liquid chromatographic method for the determination of microcystins in raw and treated waters.
Lawton AL, et al.
Analyst, 119(7), 1525-1530 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service