Skip to Content
MilliporeSigma
All Photos(1)

Documents

33626

Sigma-Aldrich

Sulfanilamide

puriss. p.a., ≥98% (calc. to the dried substance)

Synonym(s):

p-Aminobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
Beilstein:
511852
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39093202
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

Assay

≥98% (calc. to the dried substance)

form

solid

impurities

≤0.002% heavy metals (as Pb)

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying, 105 °C

color

white to faint beige

mp

163-166 °C
164-166 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤200 mg/kg

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

Nc1ccc(cc1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

FDDDEECHVMSUSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfanilamide (p-Aminobenzenesulfonamide) is a para-amino substituted benzenesulfonamide. Its ortho-mono and diiodo derivatives have been synthesized.

Application

  • Study of new azo-azomethine derivatives of sulfanilamide: synthesis, characterization, spectroscopic, antimicrobial, antioxidant and anticancer activity: This study explores new derivatives of sulfanilamide synthesized through diazonium salt reactions and coupling with 2-hydroxy-3-methoxybenzaldehyde, providing insights into their potential antimicrobial, antioxidant, and anticancer activities (HS Al-Atbi, IJ Al-Assadi, et al., 2020).

Packaging

33626-100G; 33626-6X100G

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Iodination of p-Aminobenzenesulfonamide and Some Symmetrical Azobenzenesulfonamides.
Scudi JV.
Journal of the American Chemical Society, 59(8), 1480-1483 (1937)
Traci L Ness et al.
Journal of immunology (Baltimore, Md. : 1950), 173(11), 6938-6948 (2004-11-24)
CCR1 has previously been shown to play important roles in leukocyte trafficking, pathogen clearance, and the type 1/type 2 cytokine balance, although very little is known about its role in the host response during sepsis. In a cecal ligation and
Caroline A Dobbin et al.
Journal of immunology (Baltimore, Md. : 1950), 169(2), 958-965 (2002-07-05)
We propose that the 70-kDa heat shock protein (HSP70) protects virulent Toxoplasma gondii from the effects of the host by immunomodulation. This hypothesis was tested using quercetin and antisense oligonucleotides targeting the start codon of the virulent T. gondii HSP70
Joseph A Hollenbaugh et al.
Journal of immunology (Baltimore, Md. : 1950), 177(5), 3004-3011 (2006-08-22)
We previously reported that IFN-gamma secreted by donor cytotoxic T cell 1 (Tc1) cells was the most important factor in promoting EG7 (an OVA transfection the EL4 thymoma) rejection in mice. In this study, we show that the ability of
Edward E Knaus et al.
Bioorganic & medicinal chemistry letters, 21(19), 5892-5896 (2011-08-20)
A series of compounds incorporating regioisomeric phenylethynylbenzenesulfonamide moieties has been investigated for the inhibition of four human carbonic anhydrase (hCA, EC 4.2.1.1) isoforms, hCA I, II, IX and XII. Inhibition between the low nanomolar to the milliomolar range has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service