Skip to Content
MilliporeSigma
All Photos(1)

Documents

33499

Supelco

Dimoxystrobin

PESTANAL®, analytical standard

Synonym(s):

(E)-2-(2,5-Dimethylphenoxymethyl)-α-methoxyimino-N-methylphenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O3
CAS Number:
Molecular Weight:
326.39
Beilstein:
8430159
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)C(=N/OC)\c1ccccc1COc2cc(C)ccc2C

InChI

1S/C19H22N2O3/c1-13-9-10-14(2)17(11-13)24-12-15-7-5-6-8-16(15)18(21-23-4)19(22)20-3/h5-11H,12H2,1-4H3,(H,20,22)/b21-18-

InChI key

WXUZAHCNPWONDH-UZYVYHOESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimoxystrobin belongs to the strobilurin class of fungicides and its mode of action involves the inhibition of the mitochondrial respiration of fungus.

Application

Dimoxystrobin may be used as a reference standard for the determination of the fungicide in baby food using solid-phase microextraction, followed by gas chromatography with mass spectrometric detection (SPME-GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Method development and validation for strobilurin fungicides in baby foods by solid-phase microextraction gas chromatography-mass spectrometry.
Vinas P, et al.
Journal of Chromatography A, 1216(1), 140-146 (2019)
Sylvain Bart et al.
Ecotoxicology and environmental safety, 181, 518-524 (2019-06-25)
The use of pesticides in agroecosystems can have negative effects on earthworms, which play key roles in soil functioning such as organic matter decomposition. The aim of this study was to assess the effects of two fungicides (Cuprafor micro®, composed
Joël Amossé et al.
Ecotoxicology and environmental safety, 203, 110979-110979 (2020-07-18)
Recent EFSA (European Food Safety Authority) reports highlighted that the ecological risk assessment of pesticides needed to go further by taking more into account the impacts of chemicals on biodiversity under field conditions. We assessed the effects of two commercial
Joanna Kania et al.
Pesticide biochemistry and physiology, 148, 166-174 (2018-06-13)
Cultivation of oilseed rape requires application of specific fungicides. Besides their protective role, they can potentially influence the expression and activity of crucial enzymes in the plant. Among the large number of enzymes expressed in plants, aminopeptidases play a key
Xinyi Li et al.
Journal of separation science, 43(11), 2133-2141 (2020-02-27)
Molecularly imprinted polymers for strobilurin fungicides were prepared by precipitation polymerization employing azoxystrobin as template molecular together with methacrylic acid monomer and trimethylolpropane triacrylate cross-linker. Morphological characterization showed molecularly imprinted polymers were uniform spherical particles with about 0.2 μm in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service