Skip to Content
MilliporeSigma
All Photos(2)

Documents

32666

Supelco

Melamine-13C3

VETRANAL®, analytical standard

Synonym(s):

1,3,5-Triazine-2,4,6-triamine-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3H6N6
CAS Number:
Molecular Weight:
129.10
Beilstein:
11316300
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable

mp

>300 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+3

SMILES string

N[13c]1n[13c](N)n[13c](N)n1

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)/i1+1,2+1,3+1

InChI key

JDSHMPZPIAZGSV-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Melamine-13C3 may be used as an analytical standard for the determination of melamine in milk powder samples by liquid chromatography-electrospray ionization-isotope dilution mass spectrometry (LC-ESI-IDMS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Urinary tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of isotope-labeled analogs on the LC-IDMS measurement by comparison of ESI responses and matrix effect of melamine, 13C3-melamine, 13C3 + 15N3-melamine, and 15N3-melamine.
Li XQ, et al.
Analytical and Bioanalytical Chemistry, 409(12), 3233-3243 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service