Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08614

Sigma-Aldrich

2,4,6-Triamino-1,3,5-triazine

for synthesis

Synonym(s):

2,4,6-Triamino-1,3,5-triazine, Melamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N6
CAS Number:
Molecular Weight:
126.12
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-615-4
NACRES:
NA.22

vapor pressure

66.65 hPa ( 315 °C)

Quality Level

form

powder

autoignition temp.

>600 °C

potency

3161 mg/kg LD50, oral (Rat)
>1000 mg/kg LD50, skin (Rabbit)

pH

7-8 (20 °C, 32 g/L in H2O)

bp

>354 °C/1013 hPa (sublimed)

mp

354 °C (decomposition)

solubility

3.2 g/L

density

1.57 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

no temp limit

SMILES string

[nH]1[c]([nH][c]([nH][c]1=N)=N)=N

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)

InChI key

JDSHMPZPIAZGSV-UHFFFAOYSA-N

Application



  • Synthesis, biological and in silico studies of a tripodal schiff base derived from 2, 4, 6-triamino-1, 3, 5-triazine: Discusses the synthesis and potential applications of a new Schiff base derived from 2,4,6-triamino-1,3,5-triazine in pharmaceuticals and biotechnology (Oruma et al., 2021).



Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Urinary tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service