Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM48460

Supelco

Pesticide Surrogate Spike Mix

certified reference material, TraceCERT®, 200 μg/mL each component in acetone, ampule of 1 mL

Synonym(s):

Pesticide Surrogate Spike Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8081

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL each component in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    2,4,5,6-Tetrachloro-m-xylene
    2,2′,3,3′,4,4′,5,5′,6,6′-PCB

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Endocrine system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ernestina Perez-Gonzalez et al.
Bulletin of environmental contamination and toxicology, 98(4), 454-459 (2017-01-23)
The adverse effect of pesticides on non-target wildlife and human health is a primary concern in the world, but in Mexico, we do not know which wildlife species are at the greatest risk. The aim of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service