Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45430

Supelco

Dicamba

PESTANAL®, analytical standard

Synonym(s):

3,6-Dichloro-2-methoxybenzoic acid, 3,6-Dichloro-o-anisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(OCH3)CO2H
CAS Number:
Molecular Weight:
221.04
Beilstein:
2453039
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

112-116 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

COc1c(Cl)ccc(Cl)c1C(O)=O

InChI

1S/C8H6Cl2O3/c1-13-7-5(10)3-2-4(9)6(7)8(11)12/h2-3H,1H3,(H,11,12)

InChI key

IWEDIXLBFLAXBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dicamba is a polar phenoxy herbicide, used for the post-emergence control of broad-leaf weeds in grasslands and cereals.

Application

Dicamba may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solid-phase extraction of dicamba and picloram from water and soil samples for HPLC analysis
Krzyszowska.JA and Vance.FG
Journal of Agricultural and Food Chemistry, 42(8), 1693-1696 (1994)
Analysis of dicamba from water using solid-phase extraction and ion-pair high-performance liquid chromatography
Arjmand M
Journal of Agricultural and Food Chemistry, 36(3), 492-494 (1988)
Determination of commonly used polar herbicides in agricultural drainage waters in Australia by HPLC.
Tran A T, et al.
Chemosphere, 67(5), 944-953 (2007)
Manoj Ghaste et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-17)
Dicamba is a moderately volatile herbicide used for post-emergent control of broadleaf weeds in corn, soybean, and a number of other crops. With increased use of dicamba due to the release of dicamba-resistant cotton and soybean varieties, growing controversy over
N V González et al.
Journal of hazardous materials, 163(1), 337-343 (2008-08-05)
In the present study the cytogenetic and genotoxic effect of benzoic herbicide dicamba and its Argentinean commercial formulation banvel (57.71% dicamba) was evaluated and whether this effect is mediated through oxidative damage or not. The protective role of vitamin E

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service