Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32245

Supelco

Ochratoxin A-d5 solution

10 μg/mL in acetonitrile, analytical standard

Synonym(s):

Ochratoxin A-(phenyl-d5) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20D5H13ClNO6
CAS Number:
Molecular Weight:
408.84
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+5

storage temp.

−20°C

SMILES string

[2H]c1c([2H])c([2H])c(C[C@H](NC(=O)c2cc(Cl)c3C[C@@H](C)OC(=O)c3c2O)C(O)=O)c([2H])c1[2H]

InChI

1S/C20H18ClNO6/c1-10-7-12-14(21)9-13(17(23)16(12)20(27)28-10)18(24)22-15(19(25)26)8-11-5-3-2-4-6-11/h2-6,9-10,15,23H,7-8H2,1H3,(H,22,24)(H,25,26)/t10-,15+/m1/s1/i2D,3D,4D,5D,6D

InChI key

RWQKHEORZBHNRI-CZHKJLSSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ochratoxin A-d5 is the d5 isotope-labelled analog of mycotoxin, Ochratoxin A.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service