Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31526

Supelco

Temephos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20O6P2S3
CAS Number:
Molecular Weight:
466.47
Beilstein:
1896901
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

COP(=S)(OC)Oc1ccc(Sc2ccc(OP(=S)(OC)OC)cc2)cc1

InChI

1S/C16H20O6P2S3/c1-17-23(25,18-2)21-13-5-9-15(10-6-13)27-16-11-7-14(8-12-16)22-24(26,19-3)20-4/h5-12H,1-4H3

InChI key

WWJZWCUNLNYYAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Temephos is an organophosphate mostly used as a larvicide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Temephos may be used as standard in its identification in water samples using LC–DAD and LC–APCI.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resistance to the organophosphate temephos: mechanisms, evolution and reversion in an Aedes aegypti laboratory strain from Brazil.
Melo-Santos MA
Acta Tropica, 113(2), 180-189 (2010)
Identification of fenthion and temephos and their transformation products in water by high-performance liquid chromatography with diode array detection and atmospheric pressure chemical ionization mass spectrometric detection.
Lacorte, Si'lvia, et al.
Journal of Chromatography A, 777.1, 99-114 (1997)
Mirna Velki et al.
Chemosphere, 90(11), 2690-2696 (2013-01-10)
Despite its negative effects, formalin has been often used for the expulsion of earthworms due to its high efficiency; however it is not known whether it will affect any significant measurable molecular processes in sampled earthworms. The aim of this
Clare Strode et al.
PloS one, 7(8), e39439-e39439 (2012-08-08)
The mosquito Aedes aegypti is one of the most important disease vectors because it transmits two major arboviruses, dengue and yellow fever, which cause significant global morbidity and mortality. Chemical insecticides form the cornerstone of vector control. The organophosphate temephos
Seong Chun Chae et al.
Journal of medical entomology, 51(4), 817-823 (2014-08-15)
Culex pipiens molestus Forskal (Diptera: Culicidae) is the dominant mosquito species in septic tanks in South Korea. An assessment was made of the biological control potential of mud loaches, Misgurnus mizolepis Günther (Cypriniformes: Cobitidae), toward Cx. p. molestus larvae in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service