Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

36193

Supelco

Phenothrin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26O3
CAS Number:
Molecular Weight:
350.45
Beilstein:
8788185
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(C)=C\C1C(C(=O)OCc2cccc(Oc3ccccc3)c2)C1(C)C

InChI

1S/C23H26O3/c1-16(2)13-20-21(23(20,3)4)22(24)25-15-17-9-8-12-19(14-17)26-18-10-6-5-7-11-18/h5-14,20-21H,15H2,1-4H3

InChI key

SBNFWQZLDJGRLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peilin Wang et al.
Journal of virology, 92(16) (2018-06-15)
Lassa virus (LASV) belongs to the Mammarenavirus genus (family Arenaviridae) and causes severe hemorrhagic fever in humans. At present, there are no Food and Drug Administration (FDA)-approved drugs or vaccines specific for LASV. Here, high-throughput screening of an FDA-approved drug
Yusuke Suzuki et al.
Journal of agricultural and food chemistry, 59(18), 10182-10190 (2011-09-01)
Photodegradation profiles of the pyrethroid insecticide phenothrin on a moistened U.S. soil thin layer was investigated by using its predominant component, the 1R-trans-isomer (I), under continuous exposure to light at >290 nm from a xenon arc lamp. Its degradation was
Edith Berger-Preiss et al.
International journal of hygiene and environmental health, 209(1), 41-56 (2005-12-24)
A new "pre-embarkation" method for aircraft disinsection was investigated using two different 2% d-phenothrin containing aerosols. Five experiments in aircrafts of the type Airbus 310 (4x) and Boeing 747-400 (1x) were performed. In the absence of passengers and crew the
Valeria Nardelli et al.
Journal of agricultural and food chemistry, 66(39), 10267-10273 (2018-09-13)
A sensitive and reproducible screening analytical method is described for the determination of six pyrethroids (phenothrin, permethrin, cyfluthrin, cypermethrin, deltamethrin, fenvalerate) in egg and meat samples by gas chromatography and electron capture detection (GC-ECD). A fast cleanup procedure, based on
Ayesa Paul et al.
Journal of the American Mosquito Control Association, 21(3), 305-309 (2005-10-29)
Insecticides are the primary means to control Culex pipiens, an enzootic vector of West Nile virus, in the USA. To better understand how the evolution of resistance might impact control of this insect, we investigated the levels of resistance in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service