Key Documents
26205
Hamilton® GASTIGHT® syringe, 1700 series, PTFE luer lock
1710TLL, volume 100 μL, PTFE Luer lock (with slots), needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
material
PTFE Luer lock (with slots)
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81027
needle L
, n/a
volume
100 μL
fitting
luer lock (without slots)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been used in controlling the flow rate of the liquid samples and mobile phase during ion-pair solvent extraction performed in a microchannel fabricated in a quartz glass chip.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Pharmacology for the Prehospital Professional, 54-54 (2009)
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Integration of a microextraction system on a glass chip: ion-pair solvent extraction of Fe (II) with 4, 7-diphenyl-1, 10-phenanthrolinedisulfonic acid and tri-n-octylmethylammonium chloride.
Tokeshi, Manabu, Tomoko Minagawa, and Takehiko Kitamori.
Analytical Chemistry, 72.7, 1711-1714 (2000)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)