Key Documents
20716
Hamilton® GASTIGHT® syringe, 1700 series, luer tip
1710LT, volume 100 μL, without needle, Luer Tip, needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
material
glass body
needle size
(not included)
feature
without needle Luer Tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81001
needle L
, n/a
volume
100 μL
General description
Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It was used in calibrating the instrument used for the analysis of dissolved gases by headspace sampling GC with thermal conductivity, flame ionization and FPD detectors.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)
Analysis of dissolved gases by headspace sampling gas chromatography with column and detector switching. Preliminary results.
Sarradin, Pierre-Marie, and Jean-Claude Caprais.
Analytical Communications, 33.10, 371-373 (1996)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)