Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240559

Sigma-Aldrich

1,4-Butanediol

ReagentPlus®, ≥99%

Synonym(s):

1,4-Butylene glycol, Tetramethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1633445
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent
bp:
230 °C (lit.)

grade

reagent

Quality Level

vapor density

3.1 (vs air)

product line

ReagentPlus®

Assay

≥99%

autoignition temp.

698 °F

dilution

(for general lab use)

refractive index

n20/D 1.445 (lit.)

bp

230 °C (lit.)

mp

16 °C (lit.)

density

1.017 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Butanediol is an industrial solvent that is mainly used in the preparation of important polymers such as polyurethanes and polybutylene terephthalate (PET).

Application

1,4-Butanediol may be used in the preparation of 3-buten-1-ol and tetrahydrofuran via dehydration. It may also be used as a reducing agent for carbonyl compounds to form the corresponding alcohols via transfer hydrogenation reaction.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

273.2 °F - closed cup

Flash Point(C)

134 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and mechanism of tetrahydrofuran synthesis via 1, 4-butanediol dehydration in high-temperature water
Hunter SE, et al.
The Journal of Organic Chemistry, 71(16), 6229-6239 (2006)
Dehydration of 1,4-butanediol into 3-buten-1-ol catalyzed by ceria.
Sato S, et al.
Catalysis Communications, 5(8), 397-400 (2004)
New diol processes: 1, 3-propanediol and 1, 4-butanediol
Haas T, et al.
Applied Catalysis A: General, 280(1), 83-88 (2005)
Reduction of aldehydes and ketones by transfer hydrogenation with 1, 4-butanediol.
Maytum HC, et al.
Organic Letters, 9(21), 4387-4389 (2007)
Anders Hamberg et al.
Chemical communications (Cambridge, England), 48(80), 10013-10015 (2012-09-05)
The enzyme Candida antarctica lipase B was subjected to site directed mutagenesis suggested by molecular modelling. The selectivity for the enzyme increased towards a range of diols over their corresponding monoesters as an effect of the mutations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service