Skip to Content
MilliporeSigma
All Photos(1)

Documents

21820

Sigma-Aldrich

Carbol-Fuchsin solution according to Ziehl-Neelsen

for microscopy (Bact.)

Synonym(s):

Carbol fuchsin solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy (Bact.)

Quality Level

form

liquid

technique(s)

microbe id | staining: suitable

impurities

phenol

density

0.99 g/mL at 20 °C

εmax

9.36 at 542-552 nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

suitability

bacteria

SMILES string

Cl[H].Cc1cc(ccc1N)\C(=C2\C=CC(=N)C=C2)c3ccc(N)c(C)c3

InChI

1S/C21H21N3.ClH/c1-13-11-16(5-9-19(13)23)21(15-3-7-18(22)8-4-15)17-6-10-20(24)14(2)12-17;/h3-12,22H,23-24H2,1-2H3;1H

InChI key

YCIVZWKFFBDLTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ziehl-Neelsen staining method using Carbol-Fuchsin is a crucial diagnostic tool used for the detection of acid-fast mycobacteria in tissue sections.

Application

Carbol-Fuchsin solution according to Ziehl-Neelsen is suitable for use in staining assays for acid-resistant, rod-shaped bacteria.

Other Notes

Stain for the assay of acid-resistant rod-shaped bacteria

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Bock, ed.
Romeis Mikroskopische Technik null

Articles

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service