Skip to Content
MilliporeSigma

C4165

Carbol Fuchsin

Powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Carbol Fuchsin,

form

powder

Quality Segment

color

green to brown

solubility

ethanol: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

InChI

1S/C21H21N3.ClH/c1-13-11-16(5-9-19(13)23)21(15-3-7-18(22)8-4-15)17-6-10-20(24)14(2)12-17;/h3-12,22H,23-24H2,1-2H3;1H

InChI key

YCIVZWKFFBDLTG-UHFFFAOYSA-N

General description

Carbol Fuchsin is a basic dye which forms a yellow-brown compound in presence of acid. It is mainly required for the staining of mycobacteria. The waxy walls of mycobacteria helps in retaining the dye within cells. It can also be used for the staining of bacterial spores.

Other Notes

Mixture of basic fuchsin and phenol.


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

target_organs

Nervous system,Kidney,Liver,Skin

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library