Key Documents
21744
Hamilton® needles for LT and TLL syringes
KEL-F hub, needle size 22 ga (blunt tip), needle L 51 mm (2 in.), pkg of 6 ea
About This Item
UNSPSC Code:
41122001
material
blunt tip Kel-F needle (point style 3)
needle size
22 ga (blunt tip)
feature
KEL-F hub
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 90134
needle L
51 mm (2 in.)
compatibility
for use with Hamilton LT and TLL
Footnote
For Rheodyne injector (manufacturer equivalent part no. 7215).
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)