Skip to Content
MilliporeSigma
All Photos(2)

Documents

21000-U

Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock

1010TLL, PTFE Luer lock, volume 10 mL, needle size (not included)

Synonym(s):

Hamilton 1000 Series PTFE Luer Lock Syringe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

PTFE Luer lock

needle size

(not included)

feature

label without slots

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 81620

needle L

, n/a

volume

10 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service