Skip to Content
MilliporeSigma
All Photos(1)

Documents

21603

Sigma-Aldrich

Disperse Orange 37

for microscopy

Synonym(s):

3-[4-(2,6-Dichloro-4-nitrophenylazo)-N-ethylanilino]propionitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15Cl2N5O2
CAS Number:
Molecular Weight:
392.24
Beilstein:
6348868
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy

Quality Level

form

powder

ε (extinction coefficient)

≥250 at 415-425 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCN(CCC#N)c1ccc(cc1)\N=N\c2c(Cl)cc(cc2Cl)[N+]([O-])=O

InChI

1S/C17H15Cl2N5O2/c1-2-23(9-3-8-20)13-6-4-12(5-7-13)21-22-17-15(18)10-14(24(25)26)11-16(17)19/h4-7,10-11H,2-3,9H2,1H3/b22-21+

InChI key

KHZRTXVUEZJYNE-QURGRASLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Orange 37 is a textile dye allergens. It is a component of commercial dye formulation Dispersol Black Dye used in textile industry for dyeing synthetic fibers.

Application

Disperse Orange 37 may be used during an experimental study conducted to understand the mutagenic and carcinogenic potential of textile azo dye processing plant effluent, that is known to have an impact on drinking water source.

Other Notes

Dye standard for the assay of allergy-inducing dyes in textiles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo Otávio Alves de Lima et al.
Mutation research, 626(1-2), 53-60 (2006-10-10)
Recently a textile azo dye processing plant effluent was identified as one of the sources of mutagenic activity detected in the Cristais River, a drinking water source in Brazil [G.A. Umbuzeiro, D.A. Roubicek, C.M. Rech, M.I.Z. Sato, L.D. Claxton, Investigating
Patricia A Carneiro et al.
Journal of hazardous materials, 174(1-3), 694-699 (2009-10-27)
High performance liquid chromatography coupled to a diode array detector method was developed to detect disperse dyes in water samples over the range 0.50-35 ng, with detection limits of 0.09 ng, 0.84 ng and 0.08 ng, respectively, with good repeatability
K L Hatch et al.
Journal of the American Academy of Dermatology, 32(4), 631-639 (1995-04-01)
The literature concerning textile dye dermatitis published during the last decade was reviewed. Sixty-one cases of dye-allergic contact dermatitis in which the presentation or course of the dermatitis was unusual or the dye allergen was one not previously reported have
K.L. Hatch
Textile Res., 54, 664-664 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service