Skip to Content
MilliporeSigma
Disperse Orange 37 for microscopy

Disperse Orange 37

Disperse Orange 37 for microscopy
Synonym(s):
3-[4-(2,6-Dichloro-4-nitrophenylazo)-N-ethylanilino]propionitrile
Empirical Formula (Hill Notation):
C17H15Cl2N5O2
CAS Number:
Molecular Weight:
392.24
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing