Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

195243

Sigma-Aldrich

Naphthol Blue Black

Dye content 80 %

Synonym(s):

Acid Black 1, Amido Black 10B, Buffalo Black NBR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
Colour Index Number:
20470
Beilstein:
5374263
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 80%

solubility

water: 1 mg/mL, opaque, blue to very deep blue

λmax

618 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Nc1c(\N=N\c2ccc(cc2)[N+]([O-])=O)c(cc3cc(c(\N=N\c4ccccc4)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N6O9S2.2Na/c23-19-18-12(11-17(39(35,36)37)21(22(18)29)27-24-13-4-2-1-3-5-13)10-16(38(32,33)34)20(19)26-25-14-6-8-15(9-7-14)28(30)31;;/h1-11,29H,23H2,(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2/b26-25+,27-24+;;

InChI key

AOMZHDJXSYHPKS-DROYEMJCSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Naphthol blue black (NBB) is an textile diazo dye, with high photo- and thermal-stability. Photoelectrochemical degradation of this dye yields colorless quinone as end product. Pure grade of NBB is generally used for dyeing foodstuffs, drugs and cosmetics. It is also used as a protein stain in chromatography and electrophoresis and as a biological stain. Commercial grade of NBB is a widely used textile dye. It is used in dyeing wool, nylon, silk and in textile printing. In addition, it is also employed for coloring of soaps, anodized aluminum and casein, wood stains and writing ink preparation.

Application

Naphthol blue black (NBB) has been used to stain soluble proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmental photochemistry on semiconductor surfaces. Visible light induced degradation of a textile diazo dye, naphthol blue black, on TiO2 nanoparticles
Nasr C, et al.
The Journal of Physical Chemistry, 100(20), 8436-8442 (1996)
Synthesis of layered organic--inorganic nanohybrid material: an organic dye, naphthol blue black in magnesium--aluminum layered double hydroxide inorganic lamella
bin H, et al.
Materials Science and Engineering, B, 88(1), 98-102 (2002)
Characteristics of Acaciamangium shoot apical meristems in natural and in vitro conditions in relation to heteroblasty
Hatt, Clemence, et al.
Trees (Berlin, Germany : West), 26(3), 1031-1044 (2012)
Photoelectrochemical degradation of naphthol blue black diazo dye on WO3 film electrode
Luo J and Hepel M
Electrochimica Acta, 46(19), 2913-2922 (2001)
S Nethaji et al.
Chemosphere, 82(10), 1367-1372 (2010-12-24)
Chemically prepared activated carbon material derived from palm flower was used as adsorbent for removal of Amido Black dye in aqueous solution. Batch adsorption studies were performed for the removal of Amido Black 10B (AB10B), a di-azo acid dye from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service