Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19022

Supelco

Sodium 1-butanesulfonate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

1-Butanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SO3Na
CAS Number:
Molecular Weight:
160.17
Beilstein:
3716662
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

form

solid

quality

LiChropur

mol wt

160.17 g/mol

technique(s)

ion pair chromatography: suitable

mp

>300 °C (lit.)

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1
λ: 220 nm Amax: 0.06
λ: 230 nm Amax: 0.04
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].CCCCS([O-])(=O)=O

InChI

1S/C4H10O3S.Na/c1-2-3-4-8(5,6)7;/h2-4H2,1H3,(H,5,6,7);/q;+1/p-1

InChI key

XQCHMGAOAWZUPI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-butane sulfonate is commonly used as an anion-pairing agent in the supercritical fluid chromatography (SFC) method and reversed-phase chromatographic separation systems.

Application

Sodium 1-butanesulfonate can be used as a:
  • Buffer component in high-performance liquid chromatography (HPLC) for arsenic speciation.
  • Mobile phase component in reverse-phase column for arsenic speciation.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversed-phase liquid chromatography with mixed ion-pair reagents coupled with ICP-MS for the direct speciation analysis of selenium compounds in human urine
Zheng J, et al.
Journal of Analytical Atomic Spectrometry, 17(7), 730-735 (2002)
Development of an apple juice certified reference material for cadmium, lead, total arsenic and arsenic species
Dewi F, et al.
International Journal of Mass Spectrometry, 411, 14-20 (2016)
Dietary exposure to arsenic species in Japan in 2019 using a total diet study based on composite sample with market basket approach at the national level
Suzuki Y, et al.
Journal of food composition and analysis, 108, 104384-104384 (2022)
J Zheng et al.
Journal of chromatography. A, 874(1), 55-64 (2000-04-18)
For selenium speciation analysis, the hyphenation of chromatographic separation with element-specific detection has proved a useful technique. A powerful separation system, which is capable of resolving several biologically and environmentally important selenium compounds in a single column, is greatly needed.
M T Vincenzini et al.
Life sciences, 31(5), 463-470 (1982-08-02)
The effect of 1-butanesulfonic acid sodium salt and sodium dodecyl sulfate on the activity of highly purified and crystalline enzymes with marked differences in structure and function has been studied. The enzymes were: alcohol dehydrogenase; lactate dehydrogenase; malate dehydrogenase; isocitrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service