Skip to Content
MilliporeSigma
All Photos(1)

Documents

76955

Sigma-Aldrich

Sodium 1-pentanesulfonate monohydrate

≥98.0% (T)

Synonym(s):

1-Pentanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4SO3Na · H2O
CAS Number:
Molecular Weight:
192.21
Beilstein:
3725147
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

flakes

SMILES string

O.[Na+].CCCCCS([O-])(=O)=O

InChI

1S/C5H12O3S.Na.H2O/c1-2-3-4-5-9(6,7)8;;/h2-5H2,1H3,(H,6,7,8);;1H2/q;+1;/p-1

InChI key

FPQYXAFKHLSWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-pentanesulfonate is mainly used as an ion-pairing agent in chromatographic mobile phase.

Application


  • Application of hydrophilic interaction liquid chromatography for the quantification of succinylcholine: This study outlines a method for using sodium 1-pentanesulfonate monohydrate for ion pair chromatography in the quantification of succinylcholine (A Szterk, A Zmysłowski, S Gorinstein, 2018).

  • HPLC method development and validation for the determination of Cefaclor in human plasma: Describes the use of sodium 1-pentanesulfonate monohydrate in mobile phase preparation for the analysis of Cefaclor (S Naz, MH Shoaib, L Bashir, RI Yousuf, 2017).

  • Dissociation between urate and blood pressure in mice and in people with early Parkinson′s disease: Utilizes sodium 1-pentanesulfonate monohydrate in the mobile phase composition for analysis (X Chen, CC Umeh, RE Tainsh, DD Feng, M Maguire, 2018).

  • New liquid oral formulations of hydroxychloroquine: a physicochemical stability study: Examines stability of hydroxychloroquine formulations with sodium 1-pentanesulfonate monohydrate as part of the mobile phase (V Lebreton, B Bourcier, K Cosson, 2021).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid simultaneous determination of o-phenylphenol, diphenyl, thiabendazole, imazalil and its major metabolite in citrus fruits by liquid chromatography-mass spectrometry using atmospheric pressure photoionization.
Yoshioka N, et al.
Journal of Chromatography A, 1022(1-2), 145-150 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service