Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

181846

Supelco

Poly(dimethylsiloxane)

analytical standard, average Mw 95,000 (Typical), average Mn 50,000 (Typical)

Synonym(s):

Dimethylpolysiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Si(CH3)2O]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

5 mmHg ( 20 °C)

mol wt

average Mn 50,000 (Typical)
average Mw 95,000 (Typical)

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

SMILES string

C[Si](C)=O

InChI

1S/C2H6OSi/c1-4(2)3/h1-2H3

InChI key

SEUDSDUUJXTXSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular weight distribution of poly (dimethylsiloxane) by combining matrix-assisted laser desorption/ionization time-of-flight mass spectrometry with gel-permeation chromatography fractionation.
Montaudo, Giorgio, et al.
Rapid Communications in Mass Spectrometry, 9.12, 1158-1163 (1995)
Robust and biodegradable elastomers based on corn starch and polydimethylsiloxane (PDMS).
Ceseracciu L
ACS Applied Materials & Interfaces, 7(6), 3742-3753 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service