Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17171

Millipore

Mineral modified Glutamate Broth (Base)

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

Agency

according to ISO 16649-3:2015

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

L-arginine, 0.04 g/L
L-aspartic acid, 0.048 g/L
bromocresol purple, 0.02 g/L
calcium chloride, 0.02 g/L
L-cystine, 0.04 g/L
dipotassium phosphate, 1.8 g/L
ferric ammonium citrate, 0.02 g/L
lactose, 20 g/L
magnesium sulfate, 0.2 g/L
nicotinic acid, 0.002 g/L
pantothenic acid, 0.002 g/L
sodium formate, 0.5 g/L
thiamine, 0.002 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.7±0.2 (25 °C)

application(s)

agriculture
agriculture
environmental
food and beverages
water monitoring

microbiology

suitability

nonselective and differential for Escherichia coli
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae

Application

For the enumeration of coliform bacteria in water and waste water samples.

Preparation Note

Suspend 22.7 g of Part A and 12.7 g of Part B in 1 litre distilled water containing 5 g of ammonium chloride. Heat is necessary to dissolve the medium completely. Dispense in tubes containing inverted Durham′s tubes. Sterilize by autoclaving at 115°C for 10 minutes. For single strength medium use 11.35 g of Part A and 6.35 g of Part B in 1 litre distilled water containing 2.5 g ammonium chloride.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service