Skip to Content
MilliporeSigma

1.16122

Tryptone Bile X-glucuronide agar

ChromoCult®, ISO 16649, granular, for Escherichia coli, pkg of 500 g

Synonym(s):

TBX agar, Tryptone Bile Glucuronide medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213
Manufacturer/tradename:
ChromoCult®
Agency:
ISO 16649
Ph:
7.0-7.4 (25 °C, 36.6 g/L in H2O, after autoclaving)
Form:
granular
Suitability:
Escherichia coli
Application(s):
food and beverages
microbiology
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 16649

Quality Level

form

granular

packaging

pkg of 500 g

manufacturer/tradename

ChromoCult®

technique(s)

microbiological culture: suitable

pH

7.0-7.4 (25 °C, 36.6 g/L in H2O, after autoclaving)

solubility

31.6 g/L

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Escherichia coli

General description

Chromocult® Tryptone Bile X-glucuronide agar is a selective medium for the enumeration of Escherichia coli. The presence of the enzyme β-D-glucuronidase differentiates most E. coli from other coliforms. E. coli absorbs the chromogenic substrate 5-bromo-4-chloro-3- indolyl-β-D-glucuronide (X-β-D-glucuronide) while, the enzyme β-glucuronidase splits the bond between the chromophore 5-bromo-4-chloro-3-indolyl and the β-D-glucuronide. The formed E. coli colonies are colored blue to blue green. The growth of accompanying Gram-positive flora is largely inhibited due to the presence of bile salts and the high incubation temperature of 44°C. Some E. coli (3-4 %) are β-D-glucuronidase negative and appear as colorless colonies (e.g. most E. coli O157 strains) or do not grow at the elevated temperature of 44°C (e.g. E. coli 0157:H7).

Application

Chromocult® Tryptone Bile X-glucuronide agar is used for the enumeration, detection, and identification of β-glucuronidase positive Escherichia coli from food and feeding stuff, as well as from environmental samples in the area of food production and food handling.

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish to yellowish-brown
pH-value (25 °C): 7.0 - 7.4
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012)):
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)):
Inoculum on reference medium(Escherichia coli NCTC 13216 (WDCM00202)):
Colony count (Escherichia coli ATCC 8739 (WDCM 00012)):
Colony count (Escherichia coli ATCC 25922 (WDCM 00013)):
Colony count (Escherichia coli NCTC 13216 (WDCM00202)):
Recovery on test medium (Escherichia coli ATCC 8739 (WDCM 00012)): ≥ 50 %
Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 50 %
Recovery on test medium(Escherichia coli NCTC 13216 (WDCM00202)): ≥ 50 %
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): total inhibition
Growth (Enterococcus faecalis ATCC 29212 (WDCM 00087)): total inhibition
Growth (Citrobacter freundii ATCC 43864 (WDCM 00006)): no limit
Growth (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): no limit
Colonies (Escherichia coli ATCC 8739 (WDCM 00012)): turquoise to blue
Colonies (Escherichia coli ATCC 25922 (WDCM 00013)): turquoise to blue
Colonies ((Escherichia coli NCTC 13216 (WDCM00202)): turquoise to blue
Colonies (Citrobacter freundii ATCC 43864 (WDCM 00006)): white to green-beige
Colonies (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): white to green-beige
Incubation: 21 ± 3 hours at 44 ± 1 °C aerobic
A recovery rate of 50 % is equivalent to a productivity value of 0.5.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Tryptone Bile X-glucuronide agar


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
116122050004022536682303