Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15522

Sigma-Aldrich

D-(+)-Galactose

meets analytical specification of Ph. Eur., BP

Synonym(s):

Galactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724619
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

form

powder

Quality Level

quality

meets analytical specification of Ph. Eur., BP

technique(s)

IR spectroscopy: suitable

impurities

acid. or alk. react. impurities, in accordance
microbiological impurity, in accordance
residual solvents, in accordance
≤1.0% water (Karl Fischer)

color

white

useful pH range

5.0-7.0 (25 °C, 180 g/L)

mp

168-170 °C (lit.)

solubility

H2O: soluble 180 g/L at 20 °C

suitability

in accordance for appearance of solution
in accordance for identity (IR)

application(s)

pharmaceutical (small molecule)

SMILES string

OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6-/m0/s1

InChI key

GZCGUPFRVQAUEE-KCDKBNATSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(+)-Galactose was used for examining the specificity of staining with SBA-FITC during labeling of cells for fluorescence-activated cell sorting (FACS). It was used for selecting the transformed strains of S. cerevisiae.

Biochem/physiol Actions

Galactose is a monosaccharide sugar and is the natural antigen present on the blood cells. It induces memory loss, neurodegeneration as well as oxidative damage in mice due to systemic exposure.
Galactose is a simple monosaccharide that serves as an energy source and as an essential component of glycolipids and glycoproteins. Galactose contributes to energy metabolism via its conversion to glucose by the enzymes that constitute the Leloir pathway. Defects in the genes encoding these proteins lead to the metabolic disorder galactosemia.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth M Prescott et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(13), 8796-8801 (2002-06-22)
Transcriptional interference between genes and the regulatory elements of simple eukaryotes such as Saccharomyces cerevisiae is an unavoidable consequence of their compressed genetic arrangement. We have shown previously that with the tandem arranged genes GAL10 and GAL7, inefficient transcriptional termination
P J Simmons et al.
Blood, 78(11), 2848-2853 (1991-12-01)
Normal bone marrow cells were isolated by fluorescence-activated cell sorting (FACS) on the basis of CD34 antigen expression and then assayed in vitro for colonies of fibroblastic cells (fibroblast colony-forming units [CFU-F]). Greater than 95% of detectable CFU-F were recovered
Xu Cui et al.
Journal of neuroscience research, 84(3), 647-654 (2006-05-20)
Chronic systemic exposure of mice, rats, and Drosophila to D-galactose causes the acceleration of senescence and has been used as an aging model. The underlying mechanism is yet unclear. To investigate the mechanisms of neurodegeneration in this model, we studied
L Mortimer et al.
Mucosal immunology, 7(4), 829-841 (2013-11-21)
Entamoeba histolytica (Eh) is an extracellular protozoan parasite of the human colon, which occasionally breaches the intestinal barrier. Eradicating ameba that invades is essential for host survival. A defining but uncharacterized feature of amebic invasion is direct contact between ameba
Michelle P Christie et al.
PloS one, 9(4), e95024-e95024 (2014-04-17)
Glycosylation of biopharmaceuticals can mediate cell specific delivery by targeting carbohydrate receptors. Additionally, glycosylation can improve the physico-chemical (drug-like) properties of peptide based drug candidates. The main purpose of this study was to examine if glycosylation of the peptide enkephalin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service