Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

G5388

Sigma-Aldrich

D-(+)-Galactose

≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Galactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724619
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

product line

BioReagent

Assay

≥99% (HPLC)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

impurities

≤0.1% glucose

color

white

useful pH range

5.0-7.0 (25 °C, 180 g/L)

mp

168-170 °C (lit.)

solubility

H2O: 100 mg/mL

cation traces

Mn: ≤50 ppb

SMILES string

OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6-/m0/s1

InChI key

GZCGUPFRVQAUEE-KCDKBNATSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Galactose is a constituent of lactose. It is one of the main nutrients for newborn infants and young children.[1] It is produced in the body for the formation of lactose and is also a component of glycolipids (cerebrosides) and glycoproteins.[2]
Galactose is a constituent of lactose. It is one of the main nutrients for newborn infants and young children.[3] It is produced in the body for the formation of lactose and is also a component of glycolipids (cerebrosides) and glycoproteins.[4]

Application

D-(+)-Galactose has been used in cell line and culture medium to permits the cell growth arrest without altering the specific production rate of rh-tPA (qtPA).[5]
D-(+)-Galactose has been used in cell line and culture medium to permits the cell growth arrest without altering the specific production rate of rh-tPA (qtPA).[5]

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Initial Reactions in Anaerobic Glycolysis
Textbook of Veterinary Physiological Chemistry (2015)
Disorders of carbohydrate metabolism
Emery and Rimoin's Principles and Practice of Medical Genetics, 1-36 (2013)
Simultaneous environmental manipulations in semi-perfusion cultures of CHO cells producing rh-tPA
Vergara M, et al.
Electronic journal of Biotechnology, 15(6), 2-2 (2012)
Disorders of carbohydrate metabolism
Emery and Rimoin's Principles and Practice of Medical Genetics, 1-36 (2013)
Initial Reactions in Anaerobic Glycolysis
Textbook of Veterinary Physiological Chemistry (2015)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. Is this product plant derived?

    1 answer
    1. This product is typically derived from animal sources. The source information will be listed on the lot-specific Certificate of Origin. Please navigate to the ‘DOCUMENTATION’ section of the Product Detail Page to access a Certificate:
      https://www.sigmaaldrich.com/product/sigma/g5388#product-documentation

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    0 answers

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service