Skip to Content
MilliporeSigma
All Photos(1)

Documents

15137

Supelco

Bisphenol A bis(2,3-dihydroxypropyl) ether

analytical standard

Synonym(s):

2,2-Bis[4-(2,3-dihydroxypropoxy)phenyl]propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O6
CAS Number:
Molecular Weight:
376.44
Beilstein:
2302019
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)(c1ccc(OCC(O)CO)cc1)c2ccc(OCC(O)CO)cc2

InChI

1S/C21H28O6/c1-21(2,15-3-7-19(8-4-15)26-13-17(24)11-22)16-5-9-20(10-6-16)27-14-18(25)12-23/h3-10,17-18,22-25H,11-14H2,1-2H3

InChI key

NISVZEWKUNUGQQ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ion Mobility: TWCCSN2 value of 180.1 Å2 [M+H]+ and 185.4 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 15137 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.
Standard for determining toxic monomers released from polymers of the inner coating of cans

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographia, 34, 67-67 (1992)
Seung-Won Ahn et al.
Molecules and cells, 26(1), 74-80 (2008-06-06)
Bisphenol A bis (2,3-dihydroxypropyl) ether (BADGE.2H(2)O) is a component of commercial liquid epoxy resins commonly used in the food-packing industry and in dental sealants. There is evidence that it has significant estrogenic activity. Nur77 plays a crucial role in the
Cameron A Stewart et al.
Acta biomaterialia, 76, 283-294 (2018-06-26)
Most dental resin composite restorations are replacements for failing restorations. Degradation of the restoration-tooth margins by cariogenic bacteria results in recurrent caries, a leading cause for restoration failure. Incorporating antimicrobial agents in dental adhesives could reduce interfacial bacterial count and
Olga Pardo et al.
Journal of chromatography. A, 1107(1-2), 70-78 (2005-12-27)
A new confirmatory method for simultaneous determination of bisphenol diglycidyl ether residues (BADGE, BADGE.H(2)O, BADGE.2H(2)O, BADGE.H(2)O.HCl, BADGE.HCl, BADGE.2HCl, BFDGE and BFDGE.2HCl) from canned food has been developed. The proposed method includes extraction by pressurized liquid extraction (PLE) followed by liquid-liquid
Ans Punt et al.
Archives of toxicology, 93(7), 1941-1953 (2019-05-22)
The goal of the present study was to obtain an in vivo relevant prioritization method for the endocrine potencies of different polycarbonate monomers, by combining in vitro bioassay data with physiologically based kinetic (PBK) modelling. PBK models were developed for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service