Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

452564

Sigma-Aldrich

Bisphenol A diacetate

98%

Synonym(s):

4,4′-Isopropylidenediphenol diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2C6H4)2C(CH3)2
CAS Number:
Molecular Weight:
312.36
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

91-94 °C (lit.)

functional group

ester

SMILES string

CC(=O)Oc1ccc(cc1)C(C)(C)c2ccc(OC(C)=O)cc2

InChI

1S/C19H20O4/c1-13(20)22-17-9-5-15(6-10-17)19(3,4)16-7-11-18(12-8-16)23-14(2)21/h5-12H,1-4H3

InChI key

NSNHONPMCQYMNT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motional heterogeneity and polymorphism of bisphenol-A diacetate.
Casarini D, et al.
Magnetic Resonance in Chemistry, 31(6), 540-547 (1993)
Migaku Kawaguchi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 41-48 (2004-04-29)
A new method, based on stir bar sorptive extraction (SBSE) with in situ derivatization and thermal desorption (TD)-gas chromatography-mass spectrometry (GC-MS) is described for the determination of trace amounts of bisphenol A (BPA) in river water, urine, plasma, and saliva
Model studies and a new melt polycondensation route to poly-bisphenol a-iso/terephthalate (polyarylate).
Oishi T and Hall HK.
Journal of Polymer Science Part A: Polymer Chemistry, 30(1), 83-89 (1992)
Bassam A Sweileh et al.
Molecules (Basel, Switzerland), 15(5), 3661-3682 (2010-07-27)
This work presents a new synthetic approach to aromatic and aliphatic polycarbonates by melt polycondensation of bisphenol A diacetates with alkylene- and arylenediphenyl dicarbonates. The diphenyl dicarbonates were prepared from phenyl chloroformate and the corresponding dihydroxy compounds. The process involved
Polycarbonate synthesis by the melt phase carbonate-ester interchange reaction of bisphenol-A diacetate with dimethyl carbonate.
Deshpande MM, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 33(4), 701-705 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service