Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14079

Sigma-Aldrich

Succinic acid

puriss. p.a., ACS reagent, ≥99.5% (T)

Synonym(s):

Butanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
Beilstein:
1754069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.5% (T)

impurities

≤0.001% total nitrogen (N)

ign. residue

≤0.02%

bp

235 °C (lit.)

mp

184-186 °C (lit.)
185-191 °C

solubility

methanol: soluble 0.4 g/10 mL

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

functional group

carboxylic acid

SMILES string

OC(=O)CCC(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

KDYFGRWQOYBRFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Succinic acid may be employed in the following studies:

  • Surfactant/detergent extender/foaming agent[1]
  • As ion chelator in the electroplating process[1]
  • As a precursor for various industrially important chemicals such as adipic acid, 1,4-butanediol, tetrahydrofuran, N-methyl pyrrolidinone, 2-pyrrolidinone, succinate salts, and γ-butyrolactone[2]
  • Synthesis of biodegradable polymers such as polybutyrate succinate (PBS) and polyamides.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Lee et al.
Biotechnology and bioengineering, 72(1), 41-48 (2000-11-21)
Succinic acid was produced by fermentation of Anaerobiospirillum succiniciproducens using glycerol as a carbon source. When cells were anaerobically cultured in a medium containing 6.5 g/L glycerol, a high succinic acid yield (133%) was obtained while avoiding the formation of
Biotechnology of succinic acid production and markets for derived industrial products.
Zeikus JG, et al.
Applied Microbiology and Biotechnology, 51(5), 545-552 (1999)
Song H and Lee SY.
Enzyme and Microbial Technology, 39(3), 352-361 (2006)
Virendra Gajbhiye et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(4-5), 668-679 (2013-01-10)
The present investigation was aimed at exploring dual targeting of anti-HIV drug, zidovudine (ZDV) via sialic acid conjugated-mannosylated poly(propyleneimine) (PPI) dendritic nano-constructs. Fourth generation PPI dendrimers, sialic acid conjugated PPI dendrimers (SPPI), mannose conjugated PPI dendrimers (MPPI) and dual ligand
José Manuel Otero et al.
PloS one, 8(1), e54144-e54144 (2013-01-26)
Saccharomyces cerevisiae is the most well characterized eukaryote, the preferred microbial cell factory for the largest industrial biotechnology product (bioethanol), and a robust commerically compatible scaffold to be exploitted for diverse chemical production. Succinic acid is a highly sought after

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service