Skip to Content
MilliporeSigma
All Photos(2)

Documents

12302

Sigma-Aldrich

Ammonium iron(III) oxalate trihydrate

≥98%

Synonym(s):

Ammonium ferric oxalate, Iron ammonium oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)3[Fe(C2O4)3] · 3H2O
CAS Number:
Molecular Weight:
428.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

density

1.78 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤1000 mg/kg

SMILES string

N.N.N.O.O.O.OC(=O)C(=O)O[Fe](OC(=O)C(O)=O)OC(=O)C(O)=O

InChI

1S/3C2H2O4.Fe.3H3N.3H2O/c3*3-1(4)2(5)6;;;;;;;/h3*(H,3,4)(H,5,6);;3*1H3;3*1H2/q;;;+3;;;;;;/p-3

InChI key

LVWHKVQBJNDDEL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium ferric oxalate trihydrate decomposes completely at 275°C giving Fe2O3 and has been investigated by thermogravimetry and differential thermal analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physicochemical investigation of the decomposition products of ammonium metal carboxylates: ammonium ferric oxalate hydrate.
Hussein GAM, et al.
Journal of Analytical and Applied Pyrolysis, 31, 157-167 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service