Skip to Content
MilliporeSigma

09663

3-(Hydroxymethyl)benzonitrile

≥98.0% (GC)

Synonym(s):

3-Cyanobenzyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3235555
Assay:
≥98.0% (GC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

functional group

hydroxyl, nitrile

SMILES string

OCc1cccc(c1)C#N

InChI

1S/C8H7NO/c9-5-7-2-1-3-8(4-7)6-10/h1-4,10H,6H2

InChI key

PCOFIIVWHXIDGT-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

228.2 °F - closed cup

flash_point_c

109 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library