Select a Size
About This Item
grade
analytical standard
Quality Segment
vapor density
3.8 (vs air)
vapor pressure
1.4 mmHg ( 20 °C)
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
refractive index
n20/D 1.588 (lit.)
bp
169 °C (lit.)
mp
−15 °C (lit.)
density
1.073 g/mL at 25 °C (lit.)
application(s)
agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
Sc1ccccc1
InChI
1S/C6H6S/c7-6-4-2-1-3-5-6/h1-5,7H
InChI key
RMVRSNDYEFQCLF-UHFFFAOYSA-N
General description
Application
Legal Information
Still not finding the right product?
Explore all of our products under Thiophenol
signalword
Danger
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
122.0 °F - closed cup
flash_point_c
50 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 130974-100G | 04061838728074 |
| 8220029999 | 04022536481753 |
| 8220020100 | 04022536481746 |
| 09286-5ML-F | 04061837240140 |
| 09286-25ML-F | 04061838655356 |




