MilliporeSigma
  • Developing red-emissive ruthenium(II) complex-based luminescent probes for cellular imaging.
Developing red-emissive ruthenium(II) complex-based luminescent probes for cellular imaging.
Bioconjugate chemistry (2012-03-23)
Run Zhang, Zhiqiang Ye, Yuejiao Yin, Guilan Wang, Dayong Jin, Jingli Yuan, James A Piper
ABSTRACT
Ruthenium(II) complexes have rich photophysical attributes, which enable novel design of responsive luminescence probes to selectively quantify biochemical analytes. In this work, we developed a systematic series of Ru(II)-bipyrindine complex derivatives, [Ru(bpy)(3-n)(DNP-bpy)(n)](PF(6))(2) (n = 1, 2, 3; bpy, 2,2'-bipyridine; DNP-bpy, 4-(4-(2,4-dinitrophenoxy)phenyl)-2,2'-bipyridine), as luminescent probes for highly selective and sensitive detection of thiophenol in aqueous solutions. The specific reaction between the probes and thiophenol triggers the cleavage of the electron acceptor group, 2,4-dinitrophenyl, eliminating the photoinduced electron transfer (PET) process, so that the luminescence of on-state complexes, [Ru(bpy)(3-n)(HP-bpy)(n)](2+) (n = 1, 2, 3; HP-bpy, 4-(4-hydroxyphenyl)-2,2'-bipyridine), is turned on. We found that the complex [Ru(bpy)(DNP-bpy)(2)](2+) remarkably enhanced the on-to-off contrast ratio compared to the other two (37.8 compared to 21 and 18.7). This reveals a new strategy to obtain the best Ru(II) complex luminescence probe via the most asymmetric structure. Moreover, we demonstrated the practical utility of the complex as a cell-membrane permeable probe for quantitative luminescence imaging of the dynamic intracellular process of thiophenol in living cells. The results suggest that the new probe could be a very useful tool for luminescence imaging analysis of the toxic thiophenol in intact cells.
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%
Supelco
Thiophenol, analytical standard
Sigma-Aldrich
Thiophenol, ≥99%