Skip to Content
MilliporeSigma
All Photos(1)

Documents

09276

Sigma-Aldrich

ATX Ponceau S red staining solution

concentrate for 250 ml (6 bottles)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Quality Level

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-8-3-14(11-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,27H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23+,26-25+;;;;

InChI key

KQHKSGRIBYJYFX-WNDMICSESA-J

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Non-toxic Reagents Systems
With conventional electrophoresis reagents, there is always a risk to health. By contrast, the reagents in the ATX reagent system are non-toxic, environmentally friendly. Conventional reagents contain various toxic substances: buffer solutions contain barbituric acid, classified as a narcotic according to the Dangerous Drugs Act; staining solutions contain methanol and acetic acid which irritate the skin and airways; clarifying solutions contain methanol and dioxan isobutanol, a carcinogenic, inflammable substance which is classified as hazardous waste.
Stop using these toxic substances. For your electrophoresis, start using the non-toxic ATX reagent system and protect yourself and the environment against permanent damage.
The advantages are clear. ATX reagents are:
  • non-toxic and non-irritant to skin and airways
  • environmentally friendly
  • disposable with regular waste water
  • odorless
  • non-inflammable
  • stable at room temperature
  • specially coordinated with ATX micro-solid strips
  • available as ready-to-use solutions or as concentrates
  • guaranteed to give excellent, reproducible results

Packaging

contains 6 bottles, each for the preparation of 250 mL ready-to-use-solution

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gloria Valdés et al.
Reproductive biology and endocrinology : RB&E, 6, 13-13 (2008-03-29)
The angiogenic and invasive properties of the cytotrophoblast are crucial to provide an adequate area for feto-maternal exchange. The present study aimed at identifying the localization of interrelated angiogenic, hyperpermeability and vasodilator factors in the feto-maternal interface in pregnant guinea-pigs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service