Skip to Content
MilliporeSigma

09189

ATX Ponceau S red staining solution

BioReagent, ready-to-use solution

Synonym(s):

protein gel stain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12171500
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

BioReagent

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S([O-])(=O)=O)\N=N\c3ccc(cc3)S([O-])(=O)=O)c4ccc(cc4cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O13S4.4Na/c27-22-20(43(37,38)39)10-12-9-16(41(31,32)33)6-7-17(12)21(22)26-25-18-8-3-14(11-19(18)42(34,35)36)24-23-13-1-4-15(5-2-13)40(28,29)30;;;;/h1-11,27H,(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4/b24-23+,26-25+;;;;

InChI key

KQHKSGRIBYJYFX-WNDMICSESA-J

Features and Benefits

Non-toxic Reagents Systems
With conventional electrophoresis reagents, there is always a risk to health. By contrast, the reagents in the ATX reagent system are non-toxic, environmentally friendly. Conventional reagents contain various toxic substances: buffer solutions contain barbituric acid, classified as a narcotic according to the Dangerous Drugs Act; staining solutions contain methanol and acetic acid which irritate the skin and airways; clarifying solutions contain methanol and dioxan isobutanol, a carcinogenic, inflammable substance which is classified as hazardous waste.
Stop using these toxic substances. For your electrophoresis, start using the non-toxic ATX reagent system and protect yourself and the environment against permanent damage.
The advantages are clear. ATX reagents are:
  • non-toxic and non-irritant to skin and airways
  • environmentally friendly
  • disposable with regular waste water
  • odorless
  • non-inflammable
  • stable at room temperature
  • specially coordinated with ATX micro-solid strips
  • available as ready-to-use solutions or as concentrates
  • guaranteed to give excellent, reproducible results

Packaging

6 bottles for 1 L ready-to-use-solution


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Gloria Valdés et al.
Reproductive biology and endocrinology : RB&E, 6, 13-13 (2008-03-29)
The angiogenic and invasive properties of the cytotrophoblast are crucial to provide an adequate area for feto-maternal exchange. The present study aimed at identifying the localization of interrelated angiogenic, hyperpermeability and vasodilator factors in the feto-maternal interface in pregnant guinea-pigs.
Nowsheen Mullani et al.
Life science alliance, 4(3) (2021-01-16)
Accumulation of senescent cells is an important contributor to chronic inflammation upon aging. The inflammatory phenotype of senescent cells was previously shown to be driven by cytoplasmic DNA. Here, we propose that cytoplasmic double-stranded RNA has a similar effect. We



Global Trade Item Number

SKUGTIN
W243000-1KG-K04061837544620
W243019-5KG-K04061833902790
800238999904022536357041
66761-100MG04061832734675
W243000-10KG-K04061837544613
W243019-100G-K04061834404118
W243019-1KG-K04061834365457
W243000-SAMPLE-K04061837512452
W243000-25KG-K04061837544637
W243019-SAMPLE-K04061837512469
112372-100G04061838699961
800238025004022536357034
09189-6X1L-F04061838654182
09189-1L-F04061836968151