Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07363

Supelco

Tenivastatin ammonium salt

analytical standard

Synonym(s):

RR,1S,2S,6R,8S,8aR)-8-(2,2-Dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-1-naphthaleneheptanoic acid ammonium salt, Simvastatin acid ammonium salt, Simvastatin carboxylic acid ammonium salt, Simvastatin hydroxyl acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H40O6 NH3
CAS Number:
Molecular Weight:
453.61
Beilstein:
4836426
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C25H40O6.H3N/c1-6-25(4,5)24(30)31-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-18(26)13-19(27)14-22(28)29;/h7-8,11,15-16,18-21,23,26-27H,6,9-10,12-14H2,1-5H3,(H,28,29);1H3/t15-,16-,18+,19+,20-,21-,23-;/m0./s1

InChI key

FFPDWNBTEIXJJF-OKDJMAGBSA-N

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Guldvang et al.
Reproductive toxicology (Elmsford, N.Y.), 58, 174-183 (2015-10-18)
Endocrine modulating effects of Simvastatin (SV) and its metabolite, Simvastatin β-hydroxy acid (SVA), were investigated in H295R cells and in female Sprague-Dawley (SPRD) rats. H295R cells were exposed to SV and SVA concentrations from 0 to 10μM for 48h. Four
Ayaka Shiozawa et al.
Drug metabolism and pharmacokinetics, 36, 100364-100364 (2020-12-21)
The inhibition of CYP2C9-mediated warfarin metabolism by acid or lactone forms of statin converted in the body and effects of CYP2C9 genetic variants on their inhibition are not fully understood. Here, the effects of acid and lactone forms of statins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service