Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06270

Sigma-Aldrich

Aluminum monostearate

technical, ~75% (Al), powder

Synonym(s):

Aluminum dihydroxide stearate, Stearic acid aluminum dihydroxide salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOAl(OH)2
CAS Number:
Molecular Weight:
344.47
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

~75% (Al)

form

powder

reaction suitability

core: aluminum
reagent type: catalyst

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Al](O)O

InChI

1S/C18H36O2.Al.2H2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;;;/h2-17H2,1H3,(H,19,20);;2*1H2/q;+3;;/p-3

InChI key

UGMCXQCYOVCMTB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used for the packaging of pharmaceuticals, and in the preparation of colors for cosmetics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tarek A Ahmed et al.
AAPS PharmSciTech, 15(3), 772-780 (2014-03-22)
The objective of this study was to investigate the sustained release of a hydrophilic drug, montelukast (MK), from two biodegradable polymeric drug delivery systems, in situ implant (ISI) and in situ microparticles (ISM). N-Methyl pyrrolidone (NMP), dimethyl sulfoxide (DMSO), triacetin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service