Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05686

Millipore

DMACA Indole Disks

suitable for microbiology

Synonym(s):

Indol Detection Disks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

agriculture
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

General description

DMACA Indole disks are used for indole test to determine the ability of an organism to split tryptophan into indole and α-aminopropionic acid. The presence of indole can be detected by the addition of DMACA which results in a bluish-purple complex. With this method, it is possible to differentiate Escherichia coli from Klebsiella.

Packaging

Pack of 50 disks

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service