Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05567

Supelco

N-Acetyl-d3-glufosinate

PESTANAL®, analytical standard

Synonym(s):

2-(Acetyl-d3-amino)-4-(hydroxymethylphosphinyl)butanoic acid, Glufosinate-N-acetyl-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D3H11NO5P
CAS Number:
Molecular Weight:
226.18
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

InChI

1S/C7H14NO5P/c1-5(9)8-6(7(10)11)3-4-14(2,12)13/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)(H,12,13)/i1D3

InChI key

VZVQOWUYAAWBCP-FIBGUPNXSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service