Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31264

Supelco

3-(Methylphosphinico)propionic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9O4P
CAS Number:
Molecular Weight:
152.09
Beilstein:
2242347
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CP(O)(=O)CCC(O)=O

InChI

1S/C4H9O4P/c1-9(7,8)3-2-4(5)6/h2-3H2,1H3,(H,5,6)(H,7,8)

InChI key

QXFUBAAEKCHBQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Methylphosphinico)propionic acid is a metabolite of the pesticide, glufosinate ammonium.

Application

3-(Methylphosphinico)propionic acid may be used as reference standard in the determination of 3-(methylphosphinico)propionic acid in pesticides using gas chromatography coupled with mass spectrometry (GC/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Panseri et al.
Food chemistry, 330, 127205-127205 (2020-06-11)
The incidence of endocrine disruptors, both possible (glyphosate and glufosinate), and demonstrated (perchlorate and chlorate), was estimated in baby food commodities (meat, fish, cheese, vegetable and fruit). Ion-chromatography coupled to high resolution mass spectrometry analysis of the 105 samples did
Code of Federal Regulations: 1985-1999 (1997)
Development of a method for the simultaneous determination of phosphoric and amino acid group containing pesticides by gas chromatography with mass-selective detection: Optimization of the derivatization procedure using an experimental design approach
Stalikas.DC, et al.
Journal of Chromatography A, 872(1-2), 215-225 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service