Skip to Content
MilliporeSigma

04732

2,4-Dinitrophenylhydrazine

LiChropur, moistened with 35% water, ≥99.0% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(O2N)2C6H3NHNH2
CAS Number:
Molecular Weight:
198.14
UNSPSC Code:
41116105
NACRES:
NA.22
PubChem Substance ID:
EC Number:
204-309-3
Beilstein/REAXYS Number:
615586
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

derivatization grade ((HPLC))

Quality Segment

assay

≥99.0% (HPLC)

quality

LiChropur, moistened with 35% water

technique(s)

HPLC: suitable

mp

197-200 °C (lit.)

SMILES string

NNc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H6N4O4/c7-8-5-2-1-4(9(11)12)3-6(5)10(13)14/h1-3,8H,7H2

InChI key

HORQAOAYAYGIBM-UHFFFAOYSA-N

Application

2,4-Dinitrophenylhydrazine may be used as a derivatization agent in the analysis of aldehydes and ketones using liquid chromatography coupled with atmospheric pressure photoionization-mass spectrometry (LC-APPI-MS).

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under 2,4-Dinitrophenylhydrazine


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Desen. Expl. 1

supp_hazards

Storage Class

4.1A - Other explosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library