Skip to Content
AS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Testing
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,4-Dinitrophenylhydrazine
Linear Formula:
(O
2
N)
2
C
6
H
3
NHNH
2
CAS Number:
119-26-6
Molecular Weight:
198.14
EC Number:
204-309-3
Products
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
D199303
2,4-Dinitrophenylhydrazine,
reagent grade, 97%
Expand
View Pricing
07-5220
2,4-Dinitrophenylhydrazine,
JIS special grade
Expand
View Pricing
04732
2,4-Dinitrophenylhydrazine,
LiChropur
™
, moistened with 35% water, ≥99.0% (HPLC)
Expand
View Pricing