Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04573

Sigma-Aldrich

Thiamine hydrochloride

tested according to Ph. Eur.

Synonym(s):

Thiamio hydrochloridum, Aneurine hydrochloride, Vitamin B1 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27
Beilstein:
3851771
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
34058006
PubChem Substance ID:
NACRES:
NA.26

biological source

synthetic

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

Assay

98.5-101.0% anhydrous basis

form

solid

mp

250 °C (dec.) (lit.)

solubility

alcohol: soluble 1 gm in 100 ml (95%)
alcohol: soluble 1 gm in 315 ml (Absolute)
glycerol: soluble 1 gm in 18 ml
water: soluble 1 g/mL
benzene: insoluble
chloroform: insoluble
diethyl ether: insoluble
hexane: insoluble
methanol: very soluble
propylene glycol: soluble

storage temp.

2-8°C

SMILES string

CC1=NC(N)=C(C[N+]2=CSC(CCO)=C2C)C=N1.Cl.[Cl-]

InChI

1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1

InChI key

DPJRMOMPQZCRJU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Thiamine hydrochloride has been used as a keratin salts medium supplement for culturing log-phase fungus. It has also been used as a supplement of VB medium the minimal medium of Vogel and Bonner.

Biochem/physiol Actions

Thiamine hydrochloride is a cheap, non-toxic and efficient catalyst for the synthesis of amidoalkyl naphthols. It inhibits the production of shikonin derivatives by suspension cultures of Lithospermum erythrorhizon. Additionally, thiamine hydrochloride addition to apple juice concentrates reduces its patulin content.
Thiamine hydrochloride is a hydrochloride salt form of thiamine (Vitamin B1). It is crucial for aerobic metabolism, cell growth, transmission of nerve impulses and acetylcholine synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Fujita et al.
Plant cell reports, 1(2), 61-63 (1981-12-01)
An excellent new medium was developed for the production of shikonin derivatives by suspension cultures of Lithospermum erythrorhizon. We investigated the effects of all the components of White's medium on the production of these derivatives. Nitrate, phosphate, copper, sulfate and
Thiamine hydrochloride as an efficient catalyst for the synthesis of amidoalkyl naphthols.
Min Lei
Tetrahedron Letters, 50, 6393-6397 (2009)
Thermodynamics of (solute+ solute) and (solute+ solvent) interactions of homologous series of amino acids with thiamine hydrochloride in aqueous medium at T=(305.15, 310.15, 315.15) K: A volumetric and acoustic approach
Kumar H and Behal I
The Journal of Chemical Thermodynamics, 102, 48-62 (2016)
G Apodaca et al.
Infection and immunity, 57(10), 3081-3090 (1989-10-01)
Trichophyton rubrum is the most common dermatophyte of humans and normally colonizes the superficial layers of the epidermis (stratum corneum). Several proteinases with a possible role in the metabolism of host proteins have been purified from this fungus. The regulation
Serafettin Yazici et al.
Die Nahrung, 46(4), 256-257 (2002-09-13)
Thiamine hydrochloride, pyridoxine hydrochloride and calcium-d-pantothenate were applied apple juice concentrates (AJC) at various doses in order to reduce the patulin content. AJC samples containing high levels of patulin were stored at 22 +/- 2 degrees C and 4 degrees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service