Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77623

Sigma-Aldrich

Riboflavin Sodium Phosphate Hydrate

tested according to Ph. Eur.

Synonym(s):

Riboflavin Sodium Phosphate Hydrate, Riboflavini natrii phosphas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4NaO9P
CAS Number:
Molecular Weight:
478.33
Beilstein:
4106529
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

biological source

synthetic

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

form

powder

concentration

73-79% (RIBOFLAVIN, dry)

solubility

water: soluble 100 g/L at 20 °C

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

[Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)c2cc1C

InChI

1S/C17H21N4O9P.Na/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(25)20-17(26)19-15)5-11(22)14(24)12(23)6-30-31(27,28)29;/h3-4,11-12,14,22-24H,5-6H2,1-2H3,(H,20,25,26)(H2,27,28,29);/q;+1/p-1/t11-,12+,14-;/m0./s1

InChI key

OHSHFZJLPYLRIP-BMZHGHOISA-M

General description

Riboflavin Sodium Phosphate Hydrate is also known as flavin mononucleotide (FMN). FMN is a water-soluble micronutrient. It is enzymatically produced from riboflavin (RF). Riboflavin 5′-monophosphate is one of the constituent of enzyme cofactor flavin-adenine dinucleotide.

Application

Riboflavin Sodium Phosphate Hydrate was used as water-soluble model drug in a combined conventional inkjet printing technology with flexographic printing for the fabrication of drug delivery systems. It may be employed as initiator for the photo-initiated polymerization of acrylamide. It may be employed in chronoamperometric assay for vanadium ions.

Biochem/physiol Actions

Riboflavin Sodium Phosphate Hydrate or flavin mononucleotide (FMN) possesses fluorescence, photosensitivity and redox activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalja Genina et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 47(3), 615-623 (2012-08-21)
We combined conventional inkjet printing technology with flexographic printing to fabricate drug delivery systems with accurate doses and tailored drug release. Riboflavin sodium phosphate (RSP) and propranolol hydrochloride (PH) were used as water-soluble model drugs. Three different paper substrates: A
A 13C Nuclear-Magnetic-Resonance Study of the Enzyme Cofactor Flavin-Adenine Dinucleotide
Breitmaier E and Voelter W
European Journal of Biochemistry, 31(2), 234-238 (1972)
Study of photoluminescence property on cellulosic fabric using multifunctional biomaterials riboflavin and its derivative Flavin mononucleotide
Iyer S N, et al.
Scientific Reports, 9(1), 8696-8696 (2019)
A 13C Nuclear-Magnetic-Resonance Study of the Enzyme Cofactor Flavin-Adenine Dinucleotide.
Breitmaier E and Voelter W.
European Journal of Basic Medical Sciences, 312(2), 234-238 (1972)
Vanadium determination in water using alkaline phosphatase based screen-printed carbon electrodes modified with gold nanoparticles.
Alvarado-Gamez AL, et al.
Journal of Electroanalytical Chemistry, 693, 51-55 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service