Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

04060

Sigma-Aldrich

4-(Boc-aminomethyl)benzoic acid

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNHCH2C6H4CO2H
CAS Number:
Molecular Weight:
251.28
Beilstein:
2854286
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

white to almost white

mp

164-168 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NCc1ccc(cc1)C(O)=O

InChI

1S/C13H17NO4/c1-13(2,3)18-12(17)14-8-9-4-6-10(7-5-9)11(15)16/h4-7H,8H2,1-3H3,(H,14,17)(H,15,16)

InChI key

LNKHBRDWRIIROP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service