Skip to Content
MilliporeSigma
All Photos(1)

Documents

934283

Sigma-Aldrich

4-(Aminoethyl)-1-N-Boc-piperidine

≥95.0%

Synonym(s):

1-tert-Butoxycarbonyl-4-piperidineethanamine, 2-(N-tert-Butoxycarbonyl-4-piperidinyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24N2O2
CAS Number:
Molecular Weight:
228.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95.0%

form

semisolid

color

off-white to light yellow

storage temp.

2-8°C

SMILES string

O=C(OC(C)(C)C)N1CCC(CCN)CC1

Application

A semi-flexible linker useful for PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of PROTACs may impact degradation kinetics as well as drug design (absorption; distribution; metabolism; excretion; and toxicity; ADMET) properties of PROTACs.

Technology Spotlight:

Degrader Building Blocks for Targeted Protein Degradation

Protein Degrader Building Blocks

Legal Information

PROTAC® is a registered trademark of Arvinas Operations; Inc.; and is used under license.
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingwei Shao et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(20), e2102555-e2102555 (2021-08-17)
DNA-binding proteins, including transcription factors (TFs), play essential roles in various cellular processes and pathogenesis of diseases, deeming to be potential therapeutic targets. However, these proteins are generally considered undruggable as they lack an enzymatic catalytic site or a ligand-binding
Daniel P Bondeson et al.
Annual review of pharmacology and toxicology, 57, 107-123 (2016-10-13)
Protein homeostasis networks are highly regulated systems responsible for maintaining the health and productivity of cells. Whereas therapeutics have been developed to disrupt protein homeostasis, more recently identified techniques have been used to repurpose homeostatic networks to effect degradation of
Kedra Cyrus et al.
Molecular bioSystems, 7(2), 359-364 (2010-10-06)
Conventional genetic approaches have provided a powerful tool in the study of proteins. However, these techniques often preclude selective manipulation of temporal and spatial protein functions, which is crucial for the investigation of dynamic cellular processes. To overcome these limitations
Philipp M Cromm et al.
Cell chemical biology, 24(9), 1181-1190 (2017-06-27)
Traditional pharmaceutical drug discovery is almost exclusively focused on directly controlling protein activity to cure diseases. Modulators of protein activity, especially inhibitors, are developed and applied at high concentration to achieve maximal effects. Thereby, reduced bioavailability and off-target effects can
Momar Toure et al.
Angewandte Chemie (International ed. in English), 55(6), 1966-1973 (2016-01-13)
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service