Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03764

Sigma-Aldrich

L-β-Homoleucine hydrochloride

≥98.0%

Synonym(s):

(S)-3-Amino-5-methylhexanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO2 · HCl
CAS Number:
Molecular Weight:
181.66
Beilstein:
8073252
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

beige

application(s)

peptide synthesis

SMILES string

Cl.CC(C)C[C@H](N)CC(O)=O

InChI

1S/C7H15NO2.ClH/c1-5(2)3-6(8)4-7(9)10;/h5-6H,3-4,8H2,1-2H3,(H,9,10);1H/t6-;/m0./s1

InChI key

NXVYPYHWONGEFQ-RGMNGODLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaime A Espinoza et al.
Cell death and differentiation, 27(2), 773-789 (2019-07-10)
Pharmacological inhibition of ribosome biogenesis is a promising avenue for cancer therapy. Herein, we report a novel activity of the FDA-approved antimalarial drug amodiaquine which inhibits rRNA transcription, a rate-limiting step for ribosome biogenesis, in a dose-dependent manner. Amodiaquine triggers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service