Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02560

Supelco

4-(Dimethylamino)benzaldehyde solution

10 g/L in isopropanol, for TLC derivatization

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
Beilstein:
4132845
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

concentration

10 g/L in isopropanol

technique(s)

thin layer chromatography (TLC): suitable

SMILES string

[H]C(=O)c1ccc(cc1)N(C)C

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

spray and plunge reagent for chromatography

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H R Bruns et al.
Journal of dairy science, 98(10), 7218-7225 (2015-08-19)
Whereas most soybean feedstuffs have been extensively investigated for use in ruminant diets, a lack of information exists regarding steam-flaked soybeans (SFSB). This research evaluated various inclusion rates of SFSB in diets for lactating dairy cattle. Twelve multiparous Holstein cows
Niels Hempel et al.
Journal of applied phycology, 24(6), 1407-1418 (2012-11-06)
Microalgae are discussed as an alternative source for the production of biofuels. The lipid content compared to cultivation time of used species is the main reason for any choice of a special strain. This paper reviews more analytical data of
Md Fuad Hossain et al.
Saudi journal of biological sciences, 27(6), 1514-1520 (2020-06-04)
The present study was carried out to investigate cyanobacteria as a potential source for biodiesel production isolated from fresh water bodies of Sri Lanka. Semi mass culturing and mass culturing were carried out to obtain biomass for extracting total lipids.
Yulian Chen et al.
Asian-Australasian journal of animal sciences, 29(6), 855-864 (2016-03-10)
The objective of this study was to compare the physicochemical and nutritional properties of breast and thigh meat from commercial Chinese crossbred chickens (817 Crossbred chicken, 817C), imported commercial broilers (Arbor Acres broiler, AAB), and commercial spent hens (Hyline Brown
Li Liu et al.
PloS one, 16(2), e0247559-e0247559 (2021-02-25)
The intramuscular fat (IMF) content and fatty acid composition are important meat quality traits that are mostly affected by the cattle breed. Muscle, adipose tissue and liver are important organs involved in the development of intramuscular adipose tissue. Thus, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service